Assay Details
| Gene Symbol: Cdkn2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF474974.1 | - | 211 | 90 | ||
| AF474975.1 | - | 211 | 90 | |||
| AY679727.1 | - | 211 | 90 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Cdkn2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF474974.1 | - | 211 | 90 | ||
| AF474975.1 | - | 211 | 90 | |||
| AY679727.1 | - | 211 | 90 | |||
Set Membership: |
|