Assay Details
| Gene Symbol: H2afy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135807.1 | NP_001129279.1 | 6 - 7 | 876 | 86 | |
| GenBank mRNA | BC161981.1 | - | 6 - 7 | 876 | 9029999 | 86 |
More Information
Updated Mapping Information
Set Membership: |
|

H2afy2
Genomic Map