Assay Details
| Gene Symbol: Prl3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153736.1 | NP_714958.1 | 2 - 3 | 425 | 66 | |
| GenBank mRNA | AF526270.1 | - | 2 - 3 | 425 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prl3a1
Genomic Map