Assay Details
| Gene Symbol: Tmem14c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135169.2 | NP_001128641.1 | 1 - 2 | 92 | 118 | |
| GenBank mRNA | CB744788.1 | - | 1 - 2 | 146 | 118 | |
| FQ211234.1 | - | 1 - 2 | 82 | 118 | ||
| FQ217516.1 | - | 1 - 2 | 82 | 118 | ||
| FQ223742.1 | - | 1 - 2 | 82 | 118 | ||
| FQ224495.1 | - | 1 - 2 | 88 | 118 | ||
| FQ230083.1 | - | 1 - 2 | 110 | 118 | ||
| FQ230323.1 | - | 1 - 2 | 91 | 118 | ||
| FQ233821.1 | - | 1 - 2 | 80 | 118 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001135169.2 transcripts.
This assay is in the 5'UTR region of the NM_001135169.2 transcripts.
Set Membership: |
|

Tmem14c
Genomic Map