Assay Details
| Gene Symbol: Adtrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014144.1 | NP_001014166.1 | 4 - 5 | 618 | 67 | |
| XM_006253833.2 | XP_006253895.1 | 5 - 6 | 744 | 67 | ||
| GenBank mRNA | BC088287.1 | - | 4 - 5 | 618 | 7321485 | 67 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P165035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685314 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266369 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825697 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630061 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858027 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95733 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48163 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P165035 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P165035 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P165035 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P10594 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P111825 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 75502 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85085 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55103 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 8069 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10794591 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794597 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794590 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794596 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794593 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794598 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794594 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10794589 | - | - |
| Rat Genome 230 2.0 Array | 1379326_at | - | - |
| Rat Genome 230 2.0 Array | 1379326_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379326_at | - | - |
| Affymetrix HT Rat Focus Array | 1379326_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700002822.rn.1 | TC1700000378.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700002832.rn.1 | TC1700000378.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700002821.rn.1 | TC1700000378.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700002826.rn.1 | TC1700000378.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700002829.rn.1 | TC1700000378.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700002830.rn.1 | TC1700000378.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700002831.rn.1 | TC1700000378.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700002823.rn.1 | TC1700000378.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700002827.rn.1 | TC1700000378.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700002834.rn.1 | TC1700000378.rn.1 | RTA TC |
| Rat Expression Set 230 | 1379326_PM_at | - | - |
| Rat Expression Set 230 | 1379326_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794589 | - | - |
| Rat Genome U34 Set | rc_AA858758_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22266166 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368297 | - | - |

Adtrp
Genomic Map