Assay Details
| Gene Symbol: Eci2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006966.1 | NP_001006967.1 | 2 - 3 | 240 | 63 | |
| GenBank mRNA | BC083764.1 | - | 2 - 3 | 240 | 7193133 | 63 |
More Information
Updated Mapping Information
Set Membership: |
|

Eci2
Genomic Map