Assay Details
| Gene Symbol: Rad54b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191755.1 | NP_001178684.1 | 9 - 10 | 1749 | 112 | |
| XM_006237890.2 | XP_006237952.1 | 10 - 11 | 3554 | 112 | ||
| XM_006237891.1 | XP_006237953.1 | 8 - 9 | 3602 | 112 | ||
| XM_008763543.1 | XP_008761765.1 | 10 - 11 | 3554 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rad54b
Genomic Map