Assay Details
| Gene Symbol: Cnot11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108794.2 | NP_001102264.1 | 2 - 3 | 733 | 132 | |
| GenBank mRNA | BC158750.1 | - | 2 - 3 | 733 | 9101674 | 132 |
More Information
Updated Mapping Information
Set Membership: |
|

Cnot11
Genomic Map