Assay Details
| Gene Symbol: Fam166a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025694.2 | NP_001020865.1 | 9 - 10 | 1287 | 67 | |
| XM_008761589.1 | XP_008759811.1 | 10 - 11 | 1270 | 67 | ||
| XM_008761590.1 | XP_008759812.1 | 10 - 11 | 1262 | 67 | ||
| XM_008761591.1 | XP_008759813.1 | 9 - 10 | 1210 | 67 | ||
| XM_008761592.1 | XP_008759814.1 | 9 - 10 | 1086 | 67 | ||
| XM_008761593.1 | XP_008759815.1 | 8 - 9 | 1125 | 67 | ||
| XM_008761594.1 | XP_008759816.1 | 7 - 8 | 943 | 67 | ||
| XM_008761595.1 | XP_008759817.1 | 6 - 7 | 889 | 67 | ||
| XM_008761596.1 | XP_008759818.1 | 10 - 11 | 1340 | 67 | ||
| XM_008761597.1 | XP_008759819.1 | 10 - 11 | 1119 | 67 | ||
| GenBank mRNA | BC097396.1 | - | 9 - 10 | 1288 | 7453380 | 67 |
More Information
Updated Mapping Information
Set Membership: |
|

Fam166a
Genomic Map