Assay Details
| Gene Symbol: Ggta1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037583.1 | NP_001032672.1 | 2 - 3 | 109 | 72 | |
| XM_006234046.2 | XP_006234108.1 | 3 - 4 | 143 | 72 | ||
| GenBank mRNA | AY524048.1 | - | 2 - 3 | 109 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ggta1l1
Genomic Map