Assay Details
| Gene Symbol: Med27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106565.1 | NP_001100035.1 | 4 - 5 | 623 | 67 | |
| XM_008761638.1 | XP_008759860.1 | 4 - 5 | 629 | 67 | ||
| XM_008761639.1 | XP_008759861.1 | 3 - 4 | 662 | 67 | ||
| GenBank mRNA | BC167032.1 | - | 4 - 5 | 598 | 9092805 | 67 |
More Information
Updated Mapping Information
Set Membership: |
|

Med27
Genomic Map