Assay Details
| Gene Symbol: RGD1308742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008761800.1 | XP_008760022.1 | 1 - 2 | 142 | 127 | |
| XM_342422.5 | XP_342423.5 | 1 - 2 | 142 | 127 | ||
More Information
Set Membership: |
|

RGD1308742
Genomic Map