Assay Details
| Gene Symbol: RGD1308742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006234051.2 | XP_006234113.2 | 2 - 3 | 348 | 121 | |
| XM_342422.5 | XP_342423.5 | 2 - 3 | 345 | 121 | ||
More Information
Set Membership: |
|

RGD1308742
Genomic Map