Assay Details
| Gene Symbol: RGD1308742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006234051.2 | XP_006234113.2 | 6 - 7 | 757 | 81 | |
| XM_008761799.1 | XP_008760021.1 | 4 - 5 | 523 | 81 | ||
| XM_008761800.1 | XP_008760022.1 | 4 - 5 | 520 | 81 | ||
| XM_342422.5 | XP_342423.5 | 6 - 7 | 754 | 81 | ||
More Information
Set Membership: |
|

RGD1308742
Genomic Map