Assay Details
| Gene Symbol: RGD1308742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006234051.2 | XP_006234113.2 | 3 - 4 | 505 | 68 | |
| XM_342422.5 | XP_342423.5 | 3 - 4 | 502 | 68 | ||
More Information
Set Membership: |
|

RGD1308742
Genomic Map