Assay Details
| Gene Symbol: Ccbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013164.3 | NP_001013182.3 | 4 - 5 | 448 | 95 | |
| XM_006233810.2 | XP_006233872.1 | 4 - 5 | 825 | 95 | ||
| XM_006233811.2 | XP_006233873.1 | 4 - 5 | 918 | 95 | ||
| XM_006233812.2 | XP_006233874.1 | 4 - 5 | 376 | 95 | ||
| XM_006233813.2 | XP_006233875.2 | 4 - 5 | 388 | 95 | ||
| XM_008761658.1 | XP_008759880.1 | 5 - 6 | 1131 | 95 | ||
| XM_008761659.1 | XP_008759881.1 | 5 - 6 | 1135 | 95 | ||
| GenBank mRNA | S61960.1 | - | 4 - 5 | 589 | 95 | |
| Z49696.1 | - | 5 - 6 | 597 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccbl1
Genomic Map