Assay Details
| Gene Symbol: Dtx3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109053.1 | NP_001102523.1 | 1 - 2 | 299 | 81 | |
| XM_006248414.2 | XP_006248476.1 | 1 - 2 | 217 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dtx3l
Genomic Map