Assay Details
| Gene Symbol: Mob3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108970.1 | NP_001102440.1 | 1 - 2 | 220 | 104 | |
| XM_006238001.2 | XP_006238063.1 | 1 - 2 | 233 | 104 | ||
| XM_008763634.1 | XP_008761856.1 | 1 - 2 | 241 | 104 | ||
| GenBank mRNA | FQ222144.1 | - | 1 - 2 | 178 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108970.1, XM_006238001.2, XM_008763634.1 transcripts.
Set Membership: |
|

Mob3b
Genomic Map