Assay Details
| Gene Symbol: Tbc1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107933.1 | NP_001101403.1 | 4 - 5 | 1194 | 62 | |
| XM_006238056.2 | XP_006238118.1 | 6 - 7 | 2797 | 62 | ||
| GenBank mRNA | BC168982.1 | - | 6 - 7 | 1402 | 9030533 | 62 |
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d2
Genomic Map