Assay Details
| Gene Symbol: Sit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024344.1 | NP_001019515.1 | 1 - 2 | 87 | ||
| GenBank mRNA | BC088199.1 | - | 1 - 2 | 7377221 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sit1
Genomic Map