Assay Details
| Gene Symbol: Sit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024344.1 | NP_001019515.1 | 4 - 5 | 415 | 93 | |
| GenBank mRNA | BC088199.1 | - | 4 - 5 | 415 | 7377221 | 93 |
| FQ225852.1 | - | 3 - 4 | 330 | 93 | ||
| FQ233921.1 | - | 3 - 4 | 330 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sit1
Genomic Map