Assay Details
| Gene Symbol: Klhl32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_232840.8 | XP_232840.4 | 7 - 8 | 1770 | 62 | |
| GenBank mRNA | BC087711.1 | - | 8 - 9 | 1597 | 62 | |
More Information
Set Membership: |
|

Klhl32
Genomic Map