Assay Details
| Gene Symbol: Otud6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106639.1 | NP_001100109.1 | 5 - 6 | 904 | 127 | |
| XM_006237914.2 | XP_006237976.1 | 5 - 6 | 934 | 127 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Otud6b
Genomic Map