Assay Details
| Gene Symbol: Il11ra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139116.1 | NP_620816.1 | 4 - 5 | 385 | 145 | |
| XM_006238023.2 | XP_006238085.1 | 4 - 5 | 1005 | 145 | ||
| GenBank mRNA | AF347936.1 | - | 4 - 5 | 385 | 145 | |
| BC070921.1 | - | 4 - 5 | 380 | 7105297 | 145 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il11ra1
Genomic Map