Assay Details
| Gene Symbol: Gpr126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006227647.2 | XP_006227709.1 | 20 - 21 | 3473 | 121 | |
| XM_218313.8 | XP_218313.7 | 21 - 22 | 3558 | 121 | ||
More Information
Set Membership: |
|

Gpr126
Genomic Map