Assay Details
| Gene Symbol: Nhsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002728625.4 | XP_002728671.1 | 7 - 8 | 6232 | 88 | |
| XM_002728626.4 | XP_002728672.1 | 6 - 7 | 5563 | 88 | ||
| XM_006227668.2 | XP_006227730.1 | 7 - 8 | 5695 | 88 | ||
| XM_006227670.2 | XP_006227732.1 | 7 - 8 | 4894 | 88 | ||
| XM_006227671.2 | XP_006227733.1 | 8 - 9 | 5520 | 88 | ||
More Information
Set Membership: |
|

Nhsl1
Genomic Map