Assay Details
| Gene Symbol: Lats1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134543.2 | NP_001128015.2 | 2 - 3 | 702 | 96 | |
| XM_006227614.2 | XP_006227676.1 | 1 - 2 | 1344 | 96 | ||
| XM_008758607.1 | XP_008756829.1 | 1 - 2 | 1344 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lats1
Genomic Map