Assay Details
| Gene Symbol: Nr2e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113197.1 | NP_001106668.1 | 7 - 8 | 993 | 75 | |
| GenBank mRNA | EU316216.1 | - | 7 - 8 | 993 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nr2e1
Genomic Map