Assay Details
| Gene Symbol: Cd99 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100804.1 | NP_001094274.1 | 6 - 7 | 332 | 116 | |
| GenBank mRNA | AY262357.1 | - | 6 - 7 | 321 | 116 | |
| BC168196.1 | - | 332 | 7111460 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd99
Genomic Map