Assay Details
| Gene Symbol: Ankrd23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108211.1 | NP_001101681.1 | 7 - 8 | 916 | 85 | |
| XM_006244814.2 | XP_006244876.1 | 6 - 7 | 679 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd23
Genomic Map