Assay Details
| Gene Symbol: Ankrd23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108211.1 | NP_001101681.1 | 8 - 9 | 1017 | 93 | |
| XM_006244814.2 | XP_006244876.1 | 7 - 8 | 780 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd23
Genomic Map