Assay Details
| Gene Symbol: Lix1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106214.1 | NP_001099684.1 | 5 - 6 | 994 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P089541 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P010852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534834 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951798 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114720 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P089541 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P010852 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P089541 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P010852 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P089541 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P010852 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P330584 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 64213 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 84802 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57654 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10703415 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703417 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703416 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703419 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703418 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10703414 | - | - |
| Rat Genome 230 2.0 Array | 1379505_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1379505_at | - | - |
| Affymetrix HT Rat Focus Array | 1379505_at | - | - |
| Affymetrix HT Rat Focus Array | 1379505_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100004001.rn.1 | TC0100000627.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100004000.rn.1 | TC0100000627.rn.1 | RTA TC |
| Rat Expression Set 230 | 1379505_PM_at | - | - |
| Rat Expression Set 230 | 1379505_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703414 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21785286 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359461 | - | - |

Lix1
Genomic Map