Assay Details
| Gene Symbol: Fam120b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107466.3 | NP_001100936.2 | 9 - 10 | 2710 | 60 | |
| XM_006231141.2 | XP_006231203.1 | 9 - 10 | 2839 | 60 | ||
| XM_006231142.2 | XP_006231204.1 | 8 - 9 | 2745 | 60 | ||
| XM_008758761.1 | XP_008756983.1 | 9 - 10 | 2551 | 60 | ||
| GenBank mRNA | AY724515.1 | - | 8 - 9 | 1095 | 60 | |
| FQ210918.1 | - | 9 - 10 | 2710 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P198327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296996 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619314 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3002 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P454591 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P454591 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P454591 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P454591 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P281959 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92245 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10703362 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703359 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703357 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703355 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703356 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703352 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703360 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703361 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10703351 | - | - |
| Rat Genome 230 2.0 Array | 1394986_at | - | - |
| Rat Genome 230 2.0 Array | 1377648_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1377648_at | - | - |
| Rat Genome 230 2.0 Array | 1394986_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382875_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382875_at | - | - |
| Affymetrix HT Rat Focus Array | 1377648_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1382875_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1394986_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1382875_at | - | - |
| Affymetrix HT Rat Focus Array | 1377648_at | - | - |
| Affymetrix HT Rat Focus Array | 1394986_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100003864.rn.1 | TC0100000610.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100003868.rn.1 | TC0100000610.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100003870.rn.1 | TC0100000610.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100003874.rn.1 | TC0100000610.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100003863.rn.1 | TC0100000610.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100003869.rn.1 | TC0100000610.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100003875.rn.1 | TC0100000610.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100003862.rn.1 | TC0100000610.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100003865.rn.1 | TC0100000610.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100003871.rn.1 | TC0100000610.rn.1 | RTA TC |
| Rat Expression Set 230 | 1377648_PM_at | - | - |
| Rat Expression Set 230 | 1382875_at | - | - |
| Rat Expression Set 230 | 1377648_at | - | - |
| Rat Expression Set 230 | 1394986_at | - | - |
| Rat Expression Set 230 | 1382875_PM_at | - | - |
| Rat Expression Set 230 | 1394986_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703351 | - | - |
| Rat Genome U34 Set | rc_AA859294_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368440 | - | - |

Fam120b
Genomic Map