Assay Details
| Gene Symbol: Gkap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012160.1 | NP_001012160.1 | 11 - 12 | 1262 | 95 | |
| XM_006253559.2 | XP_006253621.1 | 11 - 12 | 1117 | 95 | ||
| XM_006253561.2 | XP_006253623.1 | 10 - 11 | 1039 | 95 | ||
| GenBank mRNA | BC083717.1 | - | 11 - 12 | 1262 | 7190699 | 95 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P122812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600729 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517841 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853755 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35779 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10517 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P122812 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P122812 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P122812 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P122812 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P14355 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 83070 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 83429 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 77071 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 41283 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6925 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55166 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10793872 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793877 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793871 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793876 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793868 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793873 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793869 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793867 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793870 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793874 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793875 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10793866 | - | - |
| Rat Genome 230 2.0 Array | 1383060_at | - | - |
| Rat Genome 230 2.0 Array | 1383059_PM_a_at | - | - |
| Rat Genome 230 2.0 Array | 1379909_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383060_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1379909_at | - | - |
| Rat Genome 230 2.0 Array | 1383059_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1383060_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383059_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1379909_at | - | - |
| Affymetrix HT Rat Focus Array | 1379909_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383060_at | - | - |
| Affymetrix HT Rat Focus Array | 1383059_PM_a_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700000857.rn.1 | TC1700000109.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700000854.rn.1 | TC1700000109.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700000856.rn.1 | TC1700000109.rn.1 | RTA TC |
| Rat Expression Set 230 | 1383060_PM_at | - | - |
| Rat Expression Set 230 | 1383059_a_at | - | - |
| Rat Expression Set 230 | 1383060_at | - | - |
| Rat Expression Set 230 | 1383059_PM_a_at | - | - |
| Rat Expression Set 230 | 1379909_PM_at | - | - |
| Rat Expression Set 230 | 1379909_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10793866 | - | - |
| Rat Genome U34 Set | rc_AA963315_at | - | - |
| Rat Genome U34 Set | rc_AI227634_at | - | - |
| Rat Genome U34 Set | rc_AI179947_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21776691 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376379 | - | - |

Gkap1
Genomic Map