Assay Details
| Gene Symbol: Zfp346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107338.1 | NP_001100808.1 | 2 - 3 | 311 | 107 | |
| XM_006253628.2 | XP_006253690.1 | 2 - 3 | 361 | 107 | ||
| GenBank mRNA | FQ212045.1 | - | 2 - 3 | 352 | 107 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp346
Genomic Map