Assay Details
| Gene Symbol: Zfp346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107338.1 | NP_001100808.1 | 6 - 7 | 825 | 60 | |
| XM_006253628.2 | XP_006253690.1 | 6 - 7 | 878 | 60 | ||
| GenBank mRNA | FQ212045.1 | - | 6 - 7 | 869 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P156445 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P156438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569077 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879925 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118527 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P156445 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P156438 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P156445 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P156438 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P156438 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P156445 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P17495 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 54558 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10797349 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797350 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797347 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797346 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797348 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797345 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10797344 | - | - |
| Rat Genome 230 2.0 Array | 1390641_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392699_at | - | - |
| Rat Genome 230 2.0 Array | 1390641_at | - | - |
| Rat Genome 230 2.0 Array | 1392699_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1390641_at | - | - |
| Affymetrix HT Rat Focus Array | 1390641_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700009273.rn.1 | TC1700001333.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700009275.rn.1 | TC1700001333.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700009276.rn.1 | TC1700001333.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700009274.rn.1 | TC1700001333.rn.1 | RTA TC |
| Rat Expression Set 230 | 1390641_PM_at | - | - |
| Rat Expression Set 230 | 1390641_at | - | - |
| Rat Expression Set 230 | 1392699_at | - | - |
| Rat Expression Set 230 | 1392699_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797344 | - | - |
| Rat Genome U34 Set | rc_AI011731_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21634953 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372902 | - | - |

Zfp346
Genomic Map