Assay Details
| Gene Symbol: Camlg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053334.1 | NP_445786.1 | 2 - 3 | 705 | 72 | |
| XM_006253658.2 | XP_006253720.1 | 2 - 3 | 747 | 72 | ||
| XM_008771509.1 | XP_008769731.1 | 2 - 3 | 749 | 72 | ||
| XM_008771510.1 | XP_008769732.1 | 2 - 3 | 749 | 72 | ||
| GenBank mRNA | AF302085.1 | - | 2 - 3 | 705 | 72 | |
| BC076377.1 | - | 2 - 3 | 707 | 7110159 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Camlg
Genomic Map