Assay Details
| Gene Symbol: Tpbpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172073.1 | NP_742070.1 | 3 - 4 | 297 | 97 | |
| GenBank mRNA | AB009890.1 | - | 3 - 4 | 297 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tpbpa
Genomic Map