Assay Details
| Gene Symbol: LOC103690020 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006235962.2 | XP_006236024.1 | 14 - 15 | 2075 | 148 | |
| XM_006235963.2 | XP_006236025.1 | 1648 | 148 | |||
| XM_006235964.2 | XP_006236026.1 | 14 - 15 | 2091 | 148 | ||
| Gene Symbol: Cd36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031561.2 | NP_113749.2 | 1649 | 148 | ||
| GenBank mRNA | BC072543.1 | - | 1649 | 7105943 | 148 | |
| L19658.1 | - | 12 - 13 | 1489 | 148 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
long-chain fatty acid metabolic process
positive regulation of cell-matrix adhesion
pattern recognition receptor signaling pathway
negative regulation of systemic arterial blood pressure
fatty acid metabolic process
triglyceride metabolic process
receptor-mediated endocytosis
phagocytosis, recognition
cell adhesion
signal transduction
cell surface receptor signaling pathway
nitric oxide mediated signal transduction
response to nutrient
response to mechanical stimulus
negative regulation of gene expression
positive regulation of macrophage derived foam cell differentiation
positive regulation of cholesterol storage
response to activity
fatty acid transport
long-chain fatty acid transport
plasma membrane long-chain fatty acid transport
negative regulation of angiogenesis
fatty acid oxidation
lipid storage
cGMP-mediated signaling
positive regulation of blood coagulation
cholesterol transport
response to estradiol
positive regulation of interleukin-12 production
positive regulation of interleukin-6 production
positive regulation of tumor necrosis factor production
cellular response to insulin stimulus
plasma lipoprotein particle clearance
low-density lipoprotein particle clearance
response to drug
lipoprotein transport
negative regulation of transcription factor import into nucleus
positive regulation of I-kappaB kinase/NF-kappaB signaling
apoptotic cell clearance
positive regulation of MAPK cascade
negative regulation of growth of symbiont in host
long-chain fatty acid import
digestive tract development
positive regulation of cytokine secretion
positive regulation of peptidyl-tyrosine phosphorylation
defense response to Gram-positive bacterium
low-density lipoprotein particle mediated signaling
positive regulation of phagocytosis, engulfment
response to growth hormone
positive regulation of macrophage cytokine production
cellular response to bacterial lipopeptide
cellular response to lipopolysaccharide
cellular response to lipoteichoic acid
cellular response to hydroperoxide
regulation of removal of superoxide radicals
positive regulation of blood microparticle formation
positive regulation of reactive oxygen species metabolic process
low-density lipoprotein receptor activity
protein binding
high-density lipoprotein particle binding
lipid binding
low-density lipoprotein particle binding
transforming growth factor beta binding
thrombospondin receptor activity
oleic acid binding
lipoteichoic acid receptor activity
lipoprotein particle binding
negative regulation of transcription from RNA polymerase II promoter
long-chain fatty acid metabolic process
positive regulation of cell-matrix adhesion
pattern recognition receptor signaling pathway
negative regulation of systemic arterial blood pressure
fatty acid metabolic process
triglyceride metabolic process
receptor-mediated endocytosis
phagocytosis, recognition
cell adhesion
signal transduction
cell surface receptor signaling pathway
nitric oxide mediated signal transduction
response to nutrient
response to mechanical stimulus
negative regulation of gene expression
positive regulation of macrophage derived foam cell differentiation
positive regulation of cholesterol storage
response to activity
fatty acid transport
long-chain fatty acid transport
plasma membrane long-chain fatty acid transport
negative regulation of angiogenesis
fatty acid oxidation
lipid storage
cGMP-mediated signaling
positive regulation of blood coagulation
cholesterol transport
response to estradiol
positive regulation of interleukin-12 production
positive regulation of interleukin-6 production
positive regulation of tumor necrosis factor production
cellular response to insulin stimulus
plasma lipoprotein particle clearance
low-density lipoprotein particle clearance
response to drug
lipoprotein transport
negative regulation of transcription factor import into nucleus
positive regulation of I-kappaB kinase/NF-kappaB signaling
apoptotic cell clearance
positive regulation of MAPK cascade
negative regulation of growth of symbiont in host
long-chain fatty acid import
digestive tract development
positive regulation of cytokine secretion
positive regulation of peptidyl-tyrosine phosphorylation
defense response to Gram-positive bacterium
low-density lipoprotein particle mediated signaling
positive regulation of phagocytosis, engulfment
response to growth hormone
positive regulation of macrophage cytokine production
cellular response to bacterial lipopeptide
cellular response to lipopolysaccharide
cellular response to lipoteichoic acid
cellular response to hydroperoxide
regulation of removal of superoxide radicals
positive regulation of blood microparticle formation
positive regulation of reactive oxygen species metabolic process
low-density lipoprotein receptor activity
protein binding
high-density lipoprotein particle binding
lipid binding
low-density lipoprotein particle binding
transforming growth factor beta binding
thrombospondin receptor activity
oleic acid binding
lipoteichoic acid receptor activity
lipoprotein particle binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P113793 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P054808 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P113795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413322 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764318 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27335 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118077 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58974 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P113793 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P054808 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P113793 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P054808 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P054808 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P113793 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P297175 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700529960 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 96162 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101498 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 102424 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36241 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 4369 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23842 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 27930 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14591 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700529960 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10853252 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853242 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853243 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853253 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10940473 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853241 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853244 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853251 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10853240 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10940473 | - | - |
| Rat Genome 230 2.0 Array | 1386901_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1367689_PM_a_at | - | - |
| Rat Genome 230 2.0 Array | 1367689_a_at | - | - |
| Rat Genome 230 2.0 Array | 1386901_at | - | - |
| Affymetrix HT Rat Focus Array | 1367689_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1367689_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1386901_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1386901_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400001511.rn.1 | TC0400000195.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400001512.rn.1 | TC0400000195.rn.1 | RTA TC |
| Rat Expression Set 230 | 1367689_a_at | - | - |
| Rat Expression Set 230 | 1367689_PM_a_at | - | - |
| Rat Expression Set 230 | 1386901_PM_at | - | - |
| Rat Expression Set 230 | 1386901_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10940473 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853240 | - | - |
| Rat Genome U34 Set | AF072411_g_at | - | - |
| Rat Genome U34 Set | rc_AA799326_s_at | - | - |
| Rat Genome U34 Set | rc_AA925752_at | - | - |
| Rat Genome U34 Set | AB005743_g_at | - | - |
| Rat Genome U34 Set | AB005743_at | - | - |
| Rat Genome U34 Set | rc_AA946368_at | - | - |
| Rat Genome U34 Set | AF072411_at | - | - |
| Rat Genome U34 Set | rc_AI060301_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22165693 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361328 | - | - |
| RatRef-12 Expression BeadChip | ILMN_53809 | - | - |

LOC103690020
Genomic Map