Assay Details
| Gene Symbol: Nelfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008339.1 | NP_001008340.1 | 8 - 9 | 1102 | 92 | |
| GenBank mRNA | BC085948.1 | - | 8 - 9 | 1102 | 7130899 | 92 |
More Information
Updated Mapping Information
Set Membership: |
|

Nelfa
Genomic Map