Assay Details
| Gene Symbol: Ogdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017461.1 | NP_001017461.1 | 11 - 12 | 1592 | 83 | |
| XM_006251446.2 | XP_006251508.1 | 11 - 12 | 1708 | 83 | ||
| XM_006251447.2 | XP_006251509.1 | 11 - 12 | 1854 | 83 | ||
| XM_006251448.2 | XP_006251510.1 | 11 - 12 | 1631 | 83 | ||
| GenBank mRNA | BC083811.1 | - | 11 - 12 | 1592 | 7110182 | 83 |
| FQ222842.1 | - | 11 - 12 | 1567 | 83 | ||
| Gene Symbol: LOC103693780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008770414.1 | XP_008768636.1 | 494 | 83 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ogdh
Genomic Map