Assay Details
| Gene Symbol: Pax2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106361.1 | NP_001099831.1 | 3 - 4 | 437 | 67 | |
| XM_006231443.2 | XP_006231505.1 | 4 - 5 | 1155 | 67 | ||
| XM_008760403.1 | XP_008758625.1 | 4 - 5 | 1152 | 67 | ||
| XM_008760404.1 | XP_008758626.1 | 4 - 5 | 1155 | 67 | ||
| XM_008760405.1 | XP_008758627.1 | 4 - 5 | 1152 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pax2
Genomic Map