Assay Details
| Gene Symbol: Tmem180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014030.1 | NP_001014052.1 | 3 - 4 | 354 | 72 | |
| XM_006231506.2 | XP_006231568.1 | 3 - 4 | 905 | 72 | ||
| XM_006231507.2 | XP_006231569.1 | 3 - 4 | 589 | 72 | ||
| XM_008760432.1 | XP_008758654.1 | 5 - 6 | 1046 | 72 | ||
| GenBank mRNA | BC083852.1 | - | 3 - 4 | 354 | 7123646 | 72 |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem180
Genomic Map