Assay Details
| Gene Symbol: Cdcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106869.1 | NP_001100339.1 | 8 - 9 | 2196 | 58 | |
| XM_006244189.2 | XP_006244251.1 | 8 - 9 | 2187 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P161880 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P790155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144049 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148043 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132451 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P161880 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P790155 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P161880 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P790155 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P161880 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P790155 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P274756 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14406 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 20340 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10921115 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921111 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921114 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921110 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921113 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921116 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921112 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921108 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921109 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10921107 | - | - |
| Rat Genome 230 2.0 Array | 1376877_at | - | - |
| Rat Genome 230 2.0 Array | 1397247_at | - | - |
| Rat Genome 230 2.0 Array | 1376877_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1397247_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376877_at | - | - |
| Affymetrix HT Rat Focus Array | 1376877_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800029445.rn.1 | TC0800003626.rn.1 | RTA TC |
| Rat Expression Set 230 | 1376877_PM_at | - | - |
| Rat Expression Set 230 | 1376877_at | - | - |
| Rat Expression Set 230 | 1397247_PM_at | - | - |
| Rat Expression Set 230 | 1397247_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921107 | - | - |
| Rat Genome U34 Set | rc_AA893164_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21159047 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_2040114 | - | - |

Cdcp1
Genomic Map