Assay Details
| Gene Symbol: Qrich1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134532.1 | NP_001128004.1 | 4 - 5 | 1775 | 83 | |
| XM_006243744.2 | XP_006243806.1 | 4 - 5 | 1775 | 83 | ||
| XM_006243745.2 | XP_006243807.1 | 4 - 5 | 2626 | 83 | ||
| XM_006243746.2 | XP_006243808.1 | 4 - 5 | 2638 | 83 | ||
| XM_006243747.2 | XP_006243809.1 | 3 - 4 | 2566 | 83 | ||
| XM_006243749.2 | XP_006243811.1 | 3 - 4 | 2578 | 83 | ||
| XM_006243750.2 | XP_006243812.1 | 2 - 3 | 1366 | 83 | ||
| XM_008766506.1 | XP_008764728.1 | 3 - 4 | 2198 | 83 | ||
| XM_008766507.1 | XP_008764729.1 | 3 - 4 | 1565 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Qrich1
Genomic Map