Assay Details
| Gene Symbol: LOC102552166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008766604.1 | XP_008764826.1 | 17 - 18 | 3412 | 60 | |
| Gene Symbol: Rad54l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134520.1 | NP_001127992.1 | 3182 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P833106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244798 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108798 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693322 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215335 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145422 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P833106 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P833106 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P833106 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P833106 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10919988 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919989 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919969 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919982 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919976 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919975 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919980 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919971 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919972 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919970 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919968 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919984 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919974 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919981 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919983 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919977 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919973 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919985 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919978 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919987 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919979 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919986 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10919967 | - | - |
| Rat Genome 230 2.0 Array | 1397576_at | - | - |
| Rat Genome 230 2.0 Array | 1397576_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1376769_at | - | - |
| Rat Genome 230 2.0 Array | 1391550_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1376769_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1391550_at | - | - |
| Affymetrix HT Rat Focus Array | 1397576_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1391550_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376769_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376769_at | - | - |
| Affymetrix HT Rat Focus Array | 1397576_at | - | - |
| Affymetrix HT Rat Focus Array | 1391550_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800030596.rn.1 | TC0800003692.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800030597.rn.1 | TC0800003692.rn.1 | RTA TC |
| Rat Expression Set 230 | 1376769_PM_at | - | - |
| Rat Expression Set 230 | 1376769_at | - | - |
| Rat Expression Set 230 | 1391550_PM_at | - | - |
| Rat Expression Set 230 | 1391550_at | - | - |
| Rat Expression Set 230 | 1397576_at | - | - |
| Rat Expression Set 230 | 1397576_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10919967 | - | - |
| Rat Genome U34 Set | rc_AI072221_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21947166 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349700 | - | - |

LOC102552166
Genomic Map