Assay Details
| Gene Symbol: LOC102552166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008766604.1 | XP_008764826.1 | 8 - 9 | 1721 | 81 | |
| Gene Symbol: Rad54l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134520.1 | NP_001127992.1 | 9 - 10 | 1491 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC102552166
Genomic Map