Assay Details
| Gene Symbol: Hmgcll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276472.1 | NP_001263401.1 | 5 - 6 | 648 | 86 | |
| XM_006243415.2 | XP_006243477.1 | 5 - 6 | 1317 | 86 | ||
| GenBank mRNA | FQ211656.1 | - | 5 - 6 | 648 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hmgcll1
Genomic Map