Assay Details
| Gene Symbol: Bckdhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019267.1 | NP_062140.1 | 8 - 9 | 975 | 68 | |
| XM_006243450.2 | XP_006243512.1 | 9 - 10 | 1032 | 68 | ||
| GenBank mRNA | BC158861.1 | - | 8 - 9 | 968 | 7932093 | 68 |
| BM385109.1 | - | 3 - 4 | 68 | |||
| CB547976.1 | - | 4 - 5 | 396 | 68 | ||
| FQ213896.1 | - | 8 - 9 | 957 | 68 | ||
| FQ226240.1 | - | 8 - 9 | 963 | 68 | ||
| M94040.1 | - | 8 - 9 | 886 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bckdhb
Genomic Map