Assay Details
| Gene Symbol: Rrp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108778.1 | NP_001102248.1 | 13 - 14 | 1318 | 62 | |
| GenBank mRNA | BC158778.1 | - | 13 - 14 | 1315 | 7134304 | 62 |
| FQ226641.1 | - | 9 - 10 | 780 | 62 | ||
| FQ230405.1 | - | 13 - 14 | 1301 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rrp9
Genomic Map